ContactUsLinkComponent
Endothelial Basal Medium, MCDB 131
Endothelial Basal Medium, MCDB 131
Supplier:  US Biological
New Product
Certificates
Endothelial Basal Medium, MCDB 131
Supplier:  US Biological
New Product
Specifications
  • Sodium pyruvate:
    110 mg/L
  • Phenol red:
    12,4 mg/L
  • L-Glutamine:
    4.,4 mg/L
  • Media Base:
    Basal Medium
  • Storage temperature:
    Room temperature/4 °C
  • Shipping temperature:
    RT