ContactUsLinkComponent
Undercounter Laboratory Refrigerators, TSU
Undercounter Laboratory Refrigerators, TSU
Supplier:  Thermo Scientific
New Product
Certificates
undefined
Undercounter Laboratory Refrigerators, TSU
Supplier:  Thermo Scientific
New Product
Specifications
  • Catalog No:
  • HERATSU405SV
  • HERATSU405GV
  • Temperature:
  • 2...8 °C
  • 2...8 °C
  • Defrost:
  • Automatic
  • Automatic
  • Shelves/drawers:
  • 2 adjustable
  • 2 adjustable
  • Electrical:
  • 230 V, 50 Hz
  • 230 V, 50 Hz
  • Plug:
  • EU
  • EU
  • W×D×H ext.:
  • 52×65×83 cm
  • 52×65×83 cm