NANOCOLOR® Photometric Tube Tests, Nitrite
Supplier: MACHEREY-NAGEL
Certificates
About this item
Photometric test kits for the determination of nitrite in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Colored pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kits are evaluated in 16 mm (AD) round cuvettes. They are pre-programmed on NANOCOLOR® photometers and are also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Remark: Measuring range on NANOCOLOR VIS II
Sludge reagent: No
Digestion: No
NanOx metal: No
NanOx: No
Delivery:
Rugged box with 20 test tubes. Sufficient for 20 tests.
Specifications
- Model:MANA985068
- Platform:NANOCOLOR® tube tests
- Storage temperature:15...25 °C
- Shelf life (months):12
- Weight:314 g (Including packaging)
Specifications
Related Information
Certificates
Search by Lot NumberIFU Photometric tube tests, NANOCOLOR® Nitrite 2.pdf IFU Photometric tube tests, NANOCOLOR® Nitrite 4.pdf
Manual Photometric Tube Tests, NANOCOLOR® Nitrite 2.pdf Manual Photometric Tube Tests, NANOCOLOR® Nitrite 4.pdf
Manual
Photometric Tube Tests, NANOCOLOR® Nitrite 2.pdf Photometric Tube Tests, NANOCOLOR® Nitrite 4.pdf
Additional Product Details
Frequently Bought Together