About this item
Photometric test kits for the determination of ortho phosphate in water samples.
Ready-to-use reagents for up to 500 determinations
- Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
- Low costs per determination
- Easy dilutions for analysis of higher concentrations
The test kits are evaluated in 10 or 50 mm rectangular cuvettes. They are preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Model:MANA91878
- Platform:NANOCOLOR® standard tests
- Storage temperature:15...25 °C
- Shelf life (months):36
- Weight:619,5 g (Including packaging)
Specifications
Related Information
Certificates
Search by Lot NumberNANOCOLOR® Ortho-Phosphate Photometric Test Kits.pdf Ortho Phosphate Standard Test.pdf