NANOCOLOR® Photometric Standard Test Kit for Cobalt
Catalog # MANA91851
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Chemical method:5 -Cl -PADAB
- Pk:1 KIT
- Description:NANOCOLOR® Cobalt, test no. 1-51
- Dimensions:160×110×130 mm (including packaging)
- Model:MANA91851
- Platform:NANOCOLOR® standard tests
- Storage temperature:15…25 °C
- Shelf life (months):24
- Weight:414,94 g (including packaging)
- For unit:UV/VIS II;VIS II;Advance;500 D
- Tests per kit:250
- Range:0,002 - 0,70 mg/L (Co²⁺)
Specifications
About this item
Photometric test kit for the determination of cobalt in water samples.
Ready-to-use reagents for up to 500 determinations
- Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
- Low costs per determination
- Easy dilutions for analysis of higher concentrations
The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Delivery: Rugged box with reagents for 250 tests.