ContactUsLinkComponent
Cleanroom Masks, Exa+ 250
Cleanroom Masks, Exa+ 250
Catalog # ISOF60411
Supplier:  ISOFIELD IRELAND
New Product
Cleanroom Masks, Exa+ 250
Catalog # ISOF60411
Supplier:  ISOFIELD IRELAND
New Product
Specifications
  • Type:
    Exa+ 250 mask with earloops and clip, sterile
  • Colour:
    White
Specifications
About this item
Validated sterile cleanroom face mask with earloops and clip.
  • Large pouch-style breathing chamber
  • Extended lower breathing chamber provides excellent coverage at neck
  • Highly efficient bacterial and particle filtration
  • Gamma irradiation, min. 25 kGy
  • Particle shedding  (Helmke drum) Cat. 1
  • Sterility Assurance Level (SAL) 10⁻⁶
  • Cleanroom class: 10/ISO 4; EU GMP (A/B)
Sterile cleanroom face masks feature an ultra-wide 250 mm length with malleable nose strip.
Packaging:
Masks are packaged 1 piece per sealed inner PE bag, 50 inner bags per sealed outer PE bag, 10 outer bags per lined carton (500 pieces).