Order Entry
Czech Republic
ContactUsLinkComponent
Magnetic Stir Bar, PTFE-Coated
Magnetic Stir Bar, PTFE-Coated
Catalog # HELL332-300-VE10
Supplier:  HELLMA
Magnetic Stir Bar, PTFE-Coated
Catalog # HELL332-300-VE10
Supplier:  HELLMA

Specifications

  • Description:
    Magnetic stir bar, PTFE-coated, 3 - 4 mm Ø, 6 - 7mm L

Specifications

About this item

For use with cell types: 109.000, 109.000F, 109.004, 109.004F, 119.000, 119.000F, 119.004, and 119.004F.

  • PTFE-coated

  • Length: 6 to 7 mm
  • 3 to 4 mm Ø