ContactUsLinkComponent
PYREX® Roller Bottle
PYREX® Roller Bottle
Catalog # BIBB1528/02
Supplier:  SciLabware
PYREX® Roller Bottle
Catalog # BIBB1528/02
Supplier:  SciLabware
Specifications
  • Description:
    Roller bottle, GL 45 thread, Ø ext. 110 mm, growth area: 670 cm² (effective), 820 cm² (overall)
Specifications
About this item
Designed for use in conjunction with roller apparatus, these PYREX® roller bottles are ideal for cultivating large quantities of anchorage dependent cells.
  • Ideal for cell culture work
  • Wide neck for easy cell harvesting
  • Non-toxic polypropylene screw cap
  • Autoclavable up to 121 °C
  • Fits standard roller apparatus
PYREX® borosilicate glass construction is specially selected for surface finish and optical clarity. This bottle can be used with membrane screw caps to allow gaseous transfer.