Order Entry
Czech Republic
ContactUsLinkComponent
 
KIMBLE® KIMAX® Non-Mercury Thermometer for Pycnometer, 14 to 38 °C
  SCER15123N-2538
undefined
KIMBLE® KIMAX® Non-Mercury Thermometer for Pycnometer, 14 to 38 °C
  SCER15123N-2538

 

  • Immersion depth:
    28 mm
  • Range:
    14...38 °C

 

 

This non-mercury thermometer is designed for use in combination with a specific gravity pycnometer (10 or 25 ml).

  • Manufactured from borosilicate glass 3,3

  • Does not contain mercury
  • Features the same subdivisions, immersion depth, and precision as mercury-filled thermometers
  • Safer and more environmentally friendly than mercury-filled thermometers

Medium length standard taper ¹⁰/₁₈ thermometer with a temperature range of 14 to 38 °C. Accurate to ±0,5 °C.