Cooling Blocks, Passive Cooler
Supplier: Brand
Certificates
About this item
Keep your samples and media cool for up to three hours with these Cooling Blocks from BRAND.
ANSI/SLAS format for use in pipetting robotic systems such as the Liquid Handling Station
- Cools microcentrifuge tubes and PCR consumables
All well positions are cooled homogenously. The Cooling Blocks work as a passive heat sink which makes them quick and easy to use without an additional power supply.
Related Information
Cooling Blocks.pdf
Application Notes
Manual Cooling Block.pdf
Manual
TDS Liquid Handling Station.pdf
Datasheet
Frequently Bought Together