Plasmodium Recombinant P. falciparum MDR1 (from E. coli)
Catalog # ABCAAB224832-100
Supplier: Abcam
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Plasmodium
- Seq:1286
- Storage conditions:−80 °C
- Biological activity:Standard
- Animal-free:N
- Protein synonyms:Multidrug resistance protein|Chloroquine resistance protein|MDR1
- Protein/peptide name:P. falciparum MDR1
- Purity:>90%
- Tested applications:SDS-PAGE
Specifications
About this item
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.