Zadání objednávky
Czech Republic
ContactUsLinkComponent
Human Recombinant SSX2 (from E. coli)
Human Recombinant SSX2 (from E. coli)
Catalog # ABCAAB181904-20
Supplier:  Abcam
Marketsource
CAS Number:  
undefined
Human Recombinant SSX2 (from E. coli)
Catalog # ABCAAB181904-20
Supplier:  Abcam
Marketsource
CAS Number:  

Specifications

  • Spojení:
    Unconjugated
  • Typ proteinu/peptidu:
    Recombinant
  • Zdroj:
    E. coli
  • Druhy:
    Human
  • Sekvence:
    1154
  • Podmínky uskladnění:
    Store at –20 °C
  • Biologická aktivita:
    Standard
  • Formulář:
    Liquid
  • Bez obsahu živočichů:
    N
  • Proteinová synonyma:
    SSX2A|SSX2B|SSX2|Protein SSX2|Cancer/testis antigen 5.2|Tumor antigen HOM-MEL-40|CT5.2
  • Název proteinu/peptidu:
    SSX2
  • Čistota:
    >85%
  • Koncentrace:
    0,25 mg/ml
  • Formulace:
    pH: 8, Constituents: 10% glycerol (glycerin, glycerine), 2,4% Urea, 0,32% Tris HCl
  • Transportní teplota:
    20
  • Testované aplikace:
    SDS-PAGE

Specifications

About this item

Recombinant human SSX2 protein (denatured) is a human full length protein, in the 1 to 188 aa range, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.