Order Entry
Czech Republic
ContactUsLinkComponent
Human Interferon gamma Receptor 1 ELISA Kit (CD119)
Human Interferon gamma Receptor 1 ELISA Kit (CD119)
Catalog # ABCAAB173193-96
Supplier:  Abcam
CAS Number:  
undefined
Human Interferon gamma Receptor 1 ELISA Kit (CD119)
Catalog # ABCAAB173193-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Human Interferon gamma Receptor 1 ELISA kit (CD119)
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    96 tests

Specifications

About this item

Human Interferon gamma Receptor 1 ELISA kit (CD119) is a single-wash 90-min Simplestep used to quantify Human Interferon gamma Receptor 1 (CD119) with a sensitivity of 140 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.