Human S100A12/CGRP ELISA Kit
ABCAAB282299-1X96
: Abcam
:
- Description:Human S100A12/CGRP ELISA kit
- Tests per kit:96 tests
Human S100A12/CGRP ELISA Kit is a single-wash 90-min Simplestep used to quantify Human S100A12/CGRP with a sensitivity of 1,89 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.