Human STAT5 (Tyr694) In-Cell ELISA Kit
Catalog # ABCAAB126429-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human STAT5 (Tyr694) In-Cell ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human STAT5 (Tyr694) In-Cell ELISA Kit is Semi-quantitative ELISA for the measurement of Human STAT5 (Tyr694) production in Human in Adherent cells samples.