Mouse PPBP ELISA Kit (CXCL7)
Catalog # ABCAAB100742-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse PPBP ELISA kit (CXCL7)
- Tests per kit:96 tests
Specifications
About this item
Mouse PPBP ELISA Kit (CXCL7) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse PPBP (CXCL7) in Mouse in Plasma, Cell culture supernatant, Serum samples.