Mouse CDKN2A/p16INK4a ELISA Kit
ABCAAB230131-96
: Abcam
:
- Description:Mouse CDKN2A/p16INK4a ELISA Kit
- Tests per kit:96 tests
Mouse CDKN2A/p16INK4a ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse CDKN2A/p16INK4a with a sensitivity of 8 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.