Pig Lipocalin-2/NGAL Antibody Pair - BSA and Azide Free
Catalog # ABCAAB317515-10X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:Lipocalin-2/NGAL
- Description:Pig Lipocalin-2 / NGAL antibody pair - BSA and azide free
- Detection range:39,06 - 2500 pg/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Pig Lipocalin-2 / NGAL antibody pair - BSA and Azide free.