Complex I Rodent Profiling ELISA Kit
Catalog # ABCAAB136809-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Complex I Rodent Profiling ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Complex I Rodent Profiling ELISA Kit is a Sandwich (qualitative) ELISA kit for the measurement of Complex I Rodent Profiling in Mouse, Rat, Human in Tissue Extracts samples.