Human/Mouse Cleaved Tau (Human Asp738/Mouse Asp713) Antibody Pair - BSA and Azide Free
Catalog # ABCAAB222286-10X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:Tau
- Description:Human/Mouse Cleaved Tau (Human Asp738/Mouse Asp713) antibody pair - BSA and azide free
- Detection range:0,313 - 10 ng/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Human/Mouse Cleaved Tau (Human Asp738/Mouse Asp713) antibody pair - BSA and Azide free.