Rat IL-1 beta ELISA Kit
ABCAAB100767-96
: Abcam
:
- Description:Rat IL-1 beta ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Rat IL-1 beta ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat IL-1 beta in Rat in plasma, cell culture supernatant, serum samples.