Mouse PDK4 ELISA Kit
Katalogové číslo ABCAAB215544-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse PDK4 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Mouse PDK4 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse PDK4 with a sensitivity of 45 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair.