Rat Myoglobin ELISA Kit
Catalog # ABCAAB157739-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat Myoglobin ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Rat Myoglobin ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat Myoglobin in Rat in plasma, serum samples.