Human APOA1 (Apolipoprotein A I) Antibody Pair - BSA and Azide free
ABCAAB253628-10X96
: Abcam
:
- Description:Human APOA1 (Apolipoprotein A I) Antibody Pair - BSA and Azide free
- Tests per kit:10×96 tests
Human APOA1 (Apolipoprotein A I) Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human APOA1 (Apolipoprotein A I).