ContactUsLinkComponent
Canine Fibrinogen ELISA Kit
Canine Fibrinogen ELISA Kit
Catalog # ABCAAB108839-96
Supplier:  Abcam
CAS Number:  
undefined
Canine Fibrinogen ELISA Kit
Catalog # ABCAAB108839-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Canine Fibrinogen ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Canine Fibrinogen ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Canine Fibrinogen in Dog in Urine samples.