Human Chorionic Gonadotropin beta ELISA Kit
Katalogové číslo ABCAAB178633-96
Dodavatel: Abcam
Číslo CAS:
Specifikace
- Description:Human Chorionic Gonadotropin beta ELISA kit
- Tests per kit:96 tests
Specifikace
O tomto produktu
Human Chorionic Gonadotropin beta ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Chorionic Gonadotropin beta in Human in Citrate plasma, Heparin Plasma, Serum samples.