ContactUsLinkComponent
Mouse MSP ELISA Kit (MST1)
Mouse MSP ELISA Kit (MST1)
Catalog # ABCAAB213879-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse MSP ELISA Kit (MST1)
Catalog # ABCAAB213879-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse MSP ELISA Kit (MST1)
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse MSP ELISA Kit (MST1) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse MSP (MST1) in Mouse in Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum samples.