Mouse Eotaxin-1 ELISA Kit
Catalog # ABCAAB100680-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Eotaxin-1 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Mouse Eotaxin-1 ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Mouse Eotaxin-1 in mouse in plasma, cell culture supernatant, serum samples.