Mouse WISP1 ELISA Kit - Intracellular
Catalog # ABCAAB290979-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse WISP1 ELISA kit - Intracellular
- Tests per kit:96 tests
Specifications
About this item
Mouse WISP1 ELISA kit - Intracellular is a single-wash 90-min Simplestep used to quantify Mouse WISP1 with a sensitivity of 10,517 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair.