Rat TNF alpha ELISPOT Kits (with un-coated plates)
Catalog # ABCAAB46595-480
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat TNF alpha ELISPOT kit (with un-coated plates)
- Tests per kit:5×96 tests
Specifications
About this item
Rat TNF alpha ELISPOT Kit (with un-coated plates) is a ELISPOT kit for the sensitive detection of Rat TNF alpha production in a single cell suspension.