Mouse Leptin R ELISA Kit
Catalog # ABCAAB267584-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Leptin R ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse Leptin R ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Leptin R in Mouse in Plasma, Cell culture supernatant, Serum samples.