Order Entry
Czech Republic
ContactUsLinkComponent
Rat BMP-2 ELISA Kit
Rat BMP-2 ELISA Kit
Catalog # ABCAAB213900-96
Supplier:  Abcam
CAS Number:  
undefined
Rat BMP-2 ELISA Kit
Catalog # ABCAAB213900-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    BMP2
  • Description:
    Rat BMP-2 ELISA kit
  • Detection method:
    Colorimetric
  • Time to Results:
    3 h 30 min
  • Detection range:
    31,2 - 2000 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    2 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Rat BMP-2 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat BMP-2 in Rat in Tissue, cell culture supernatant, Serum samples.