Mouse CXCL9 ELISA Kit
ABCAAB100725-96
: Abcam
:
- Description:Mouse CXCL9 ELISA Kit
- Tests per kit:96 tests
Mouse CXCL9 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse CXCL9 in Mouse in Plasma, Cell culture supernatant, Serum samples.