Rat CXCL7 ELISA Kit (PPBP)
Catalog # ABCAAB100781-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat CXCL7 ELISA kit (PPBP)
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Rat CXCL7 ELISA kit (PPBP) is a Sandwich (quantitative) ELISA kit for the measurement of Rat CXCL7 (PPBP) in Rat in plasma, cell culture supernatant, serum samples.