ContactUsLinkComponent
15-Deoxy-delta12,14-PGJ2 ELISA Kit
15-Deoxy-delta12,14-PGJ2 ELISA Kit
Catalog # ABCAAB133031-96
Supplier:  Abcam
CAS Number:  
undefined
15-Deoxy-delta12,14-PGJ2 ELISA Kit
Catalog # ABCAAB133031-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    15-Deoxy-delta12,14-PGJ2 ELISA kit
Specifications
About this item
15-deoxy-delta12,14-PGJ2 ELISA kit is a Competitive ELISA kit for the measurement of 15-deoxy-delta12,14-PGJ2 in Human, Pig in Saliva, urine, plasma, cell culture supernatant samples.