ContactUsLinkComponent
HLA-DPB1 ELISA Kit
Catalog # ABCAAB300336-96
Supplier:  Abcam
CAS Number:  
undefined
HLA-DPB1 ELISA Kit
Catalog # ABCAAB300336-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    HLA-DPB1 ELISA Kit
  • Tests per kit:
    96 tests
Specifications
About this item
HLA-DPB1 ELISA Kit is a single-wash 90-min Simplestep used to quantify HLA-DPB1 with a sensitivity of 8,384 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.