Tetracyclines ELISA Kit
ABCAAB285261-96
: Abcam
:
- Description:Tetracyclines ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Tetracyclines ELISA kit is a Competitive ELISA kit for the measurement of Tetracyclines in tissue, milk samples.