ContactUsLinkComponent
Mouse IL-27a ELISA Kit (Interleukin 27 p28 subunit)
Mouse IL-27a ELISA Kit (Interleukin 27 p28 subunit)
Catalog # ABCAAB206306-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse IL-27a ELISA Kit (Interleukin 27 p28 subunit)
Catalog # ABCAAB206306-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse IL-27a ELISA Kit (Interleukin 27 p28 subunit)
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse IL-27a ELISA Kit (Interleukin 27 p28 subunit) is a single-wash 90-min Simplestep used to quantify Mouse IL-27a (Interleukin 27 p28 subunit) with a sensitivity of 7,1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.