Human Cardiotrophin 1 ELISA Kit (CTF1)
Catalog # ABCAAB99996-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Cardiotrophin 1 ELISA kit (CTF1)
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Human Cardiotrophin 1 ELISA kit (CTF1) is a Sandwich (quantitative) ELISA kit for the measurement of Human Cardiotrophin 1 (CTF1) in Human in plasma, cell culture supernatant samples.