Phospho-MARCKS (S152/156) and Total MARCKS ELISA Kit
Catalog # ABCAAB279857-1X96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Pre-coated
- Target protein:MARCKS
- Description:Phospho-MARCKS (S152/156) and Total MARCKS ELISA kit
- Sample type:Cell lysate
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Phospho-MARCKS (S152/156) and Total MARCKS ELISA kit is a Semi-quantitative ELISA kit for the measurement of Phospho-MARCKS (S152/156) and Total MARCKS in Human in Cell Lysate samples.