ContactUsLinkComponent
Mouse GM-CSF ELISA Kit (CSF2)
Mouse GM-CSF ELISA Kit (CSF2)
Catalog # ABCAAB201276-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse GM-CSF ELISA Kit (CSF2)
Catalog # ABCAAB201276-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse GM-CSF ELISA Kit (CSF2)
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse GM-CSF ELISA Kit (CSF2) is a single-wash 90-min Simplestep used to quantify Mouse GM-CSF (CSF2) with a sensitivity of 3,1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.