ContactUsLinkComponent
Mouse Interferon gamma ELISA Set (without plates)
Mouse Interferon gamma ELISA Set (without plates)
  ABCAAB47618-480
 :  Abcam
 :  
undefined
Mouse Interferon gamma ELISA Set (without plates)
  ABCAAB47618-480
 :  Abcam
 :  
  • Description:
    Mouse Interferon gamma ELISA Set (without plates)
  • Seq:
    1
Mouse Interferon gamma ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Interferon gamma ELISA Set (without plates) in Mouse in plasma, cell culture supernatant, serum samples.