Mouse DLL1 ELISA Kit
Catalog # ABCAAB208342-96
Supplier: Abcam
CAS Number:
Specifications
- Doba trvání testu:One step
- Typ testu:Sandwich
- Formát:Pre-coated
- Cíl:DLL1
- Popis:Mouse DLL1 ELISA kit
- Typ vzorku:Cell culture supernatant, plasma, serum
- Detekční metoda:Colorimetric
- Doba do výsledků:1 h 30 min
- Detekční rozsah:62,5 - 4000 pg/ml
- Skladovací teplota:4 °C
- Citlivost:24 pg/ml
- Regulační status:RUO
- Testů v kitu:96 tests
Specifications
About this item
Mouse DLL1 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse DLL1 with a sensitivity of 24 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.