Human AIF ELISA Kit (Apoptosis-Inducing Factor 1)
Catalog # ABCAAB184858-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human AIF ELISA kit (Apoptosis-Inducing Factor 1)
- Tests per kit:96 tests
Specifications
About this item
Human AIF ELISA Kit (Apoptosis-Inducing Factor 1) is a single-wash 90-min Simplestep used to quantify Human AIF (Apoptosis-Inducing Factor 1) with a sensitivity of 60 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.