Pig IgM ELISA Kit
Catalog # ABCAAB190537-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Pig IgM ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Pig IgM ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Pig IgM in Pig in Plasma, serum samples.
Certifications: RUO