Order Entry
Czech Republic
ContactUsLinkComponent
p38 MAPK alpha ELISA Kit
p38 MAPK alpha ELISA Kit
Catalog # ABCAAB221012-96
Supplier:  Abcam
CAS Number:  
undefined
p38 MAPK alpha ELISA Kit
Catalog # ABCAAB221012-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Format:
    Pre-coated
  • Target protein:
    p38
  • Description:
    p38 MAPK alpha ELISA kit
  • Sample type:
    Tissue homogenate, cell lysate
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 1 min
  • Detection range:
    0,781 - 100 ng/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    300 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

p38 MAPK alpha ELISA kit is a single-wash 90-min Simplestep used to quantify p38 MAPK alpha with a sensitivity of 300 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader .