Rat IL-10 ELISA Kit
ABCAAB214566-96
: Abcam
:
- Description:Rat IL-10 ELISA Kit
- Tests per kit:96 tests
Rat IL-10 ELISA Kit is a single-wash 90-min Simplestep used to quantify Rat IL-10 with a sensitivity of 4,89 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader. - Cited in over 35 citations.