Order Entry
Czech Republic
ContactUsLinkComponent
TMB ELISA Substrate (Highest Sensitivity)
TMB ELISA Substrate (Highest Sensitivity)
Supplier:  Abcam
Certificates
undefined
TMB ELISA Substrate (Highest Sensitivity)
Supplier:  Abcam

Specifications

  • Assay duration:
    Multiple steps
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Species independent
  • Storage temperature:
    Store at 4 °C
  • Regulatory status:
    RUO

Specifications

Frequently Bought Together