Mouse IL-1 RL1 ELISA Kit (ST2)
ABCAAB213871-96
: Abcam
:
- Description:Mouse IL-1 RL1 ELISA kit (ST2)
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Mouse IL-1 RL1 ELISA kit (ST2) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IL-1 RL1 (ST2) in Mouse in cell lysate, cell culture supernatant, EDTA plasma, heparin plasma, serum samples.