Mouse CD62P ELISA Kit (P-Selectin)
Catalog # ABCAAB100737-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse CD62P ELISA Kit (P-Selectin)
- Tests per kit:96 tests
Specifications
About this item
Mouse CD62P ELISA Kit (P-Selectin) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse CD62P (P-Selectin) in Mouse in Plasma, Cell culture supernatant, Serum samples.